Learn More
CPC Scientific H-Leu-Pro-Arg-Leu-Leu-His-Thr-Asp-Ser-Arg-Met-Ala-Thr-Ile-Asp-Phe-Pro-Lys-Lys-Asp-Pro-Thr-Thr-Ser-Leu-Gly-Arg-Pro-Phe-Phe-Leu-Phe-Arg-Pro-Arg-Asn-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific NEUP008A
Encompass
A neuromedin U-related neuropeptide specifically expressed in the suprachiasmatic nuclei (SCN) of the hypothalamus. rNMS is implicated in the regulation of circadian rhythms through autocrine and/or paracrine actions. rNMS showed potent anorexigenic effects and counteracted neuropeptide Y, ghrelin and agouti-related protein-induced food intake. This compound is distributed through Peptide Institute, Inc. under the license of National Cardiovascular Center and Takeda Pharmaceutical Company LimitedONE-LETTER SEQUENCE: LPRLLHTDSRMATIDFPKKDPTTSLGRPFFLFRPRN-NH2MOLECULAR FORMULA: C193H307N57O49S1MOLECULAR WEIGHT:4241.98STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [843782-19-4], RESEARCH AREA: ObesityREFERENCES: T.Ida et al., Endocrinology, 146, 4217 (2005); K.Mori et al., EMBO J., 24, 325 (2005)
Catalog No. 50-210-9894
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.