Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neuromedin U Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578344
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578344 Supplier Invitrogen™ Supplier No. PA578344
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: A431, HeLa Predicted Reactivity: Bovine (80%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the neuromedin family of neuropeptides. The encoded protein is a precursor that is proteolytically processed to generate a biologically active neuropeptide that plays a role in pain, stress, immune-mediated inflammatory diseases and feeding regulation. Increased expression of this gene was observed in renal, pancreatic and lung cancers. Alternative splicing results in multiple transcript variants encoding different isoforms. Some of these isoforms may undergo similar processing to generate the mature peptide.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuromedin U
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene NMU
Gene Accession No. P48645
Gene Alias Neuromedin precursor-related peptide 33; Neuromedin precursor-related peptide 36; neuromedin U; neuromedin-U; Neuromedin-U-23; Neuromedin-U-25; Nmu; NmU 25; NmU-23; NmU-25; NURP33; NURP36; prepro-NMU
Gene Symbols NMU
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NMU. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10874
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.