Learn More
Diagnostic Biosystems NEURON SPECIFIC ENOLASE Neuron Specific Enolase
Supplier:  Diagnostic Biosystems RP05401
Encompass_Preferred
This antibody reacts with human isoenzymes of neuron specific enolase containing γ subunits. This antibody gives a specific staining in neurons of the brain, in islets of Langerhansin pancreas and in peripheral nerves of the skin. No staining with other tissue types has been seen.
Catalog No. 50-238-5495
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.