Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuronal Pentraxin 2 Antibody (569706) [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. FAB7816A
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7816A 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FAB7816A Supplier R&D Systems Supplier No. FAB7816A

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Neuronal Pentraxin 2 Monoclonal antibody specifically detects Neuronal Pentraxin 2 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry

Specifications

Antigen Neuronal Pentraxin 2
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 569706
Conjugate APC
Formulation PBS
Gene Alias NARP, neuronal activity-regulated pentaxin, neuronal pentraxin IIapexin, neuronal pentraxin-2, NP2, NP-II
Host Species Mouse
Immunogen Mouse myeloma cell line, NS0-derived human Neuronal Pentraxin 2, Gly16-Leu431, Accession # P47972
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4885
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.