Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuronal Pentraxin R/NPTXR Antibody (12) [Alexa Fluor™ 594], Novus Biologicals™
SDP

Catalog No. NB451250
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB451250 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB451250 Supplier R&D Systems Supplier No. NBP326911AF594

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Neuronal Pentraxin R/NPTXR Monoclonal specifically detects Neuronal Pentraxin R/NPTXR in Human samples. It is validated for ELISA.

Specifications

Antigen Neuronal Pentraxin R/NPTXR
Applications ELISA
Classification Monoclonal
Clone 12
Conjugate Alexa Fluor 594
Formulation 50 mM Sodium Borate
Gene Alias neuronal pentraxin receptor, NPR
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Neuronal Pentraxin R/NPTXR (RefSeq#: NP_055108.2; Ala24-Ala500).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23467
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.