Learn More
CPC Scientific H-Pro-Gln-Arg-Phe-NH2 (acetate salt) 25MG
SDP
Supplier:  CPC Scientific MMNP002B
Encompass
SEQUENCE: H-Pro-Gln-Arg-Phe-NH2 (acetate salt)ONE-LETTER SEQUENCE: PQRF-NH2MOLECULAR FORMULA: C25H39N9O5MOLECULAR WEIGHT: 545.64STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [152050-35-6]SYNONYMS: RESEARCH AREA: PainREFERENCES:N.A.Elshourbagy et al., J. Biol. Chem., 275, 25965 (2000)S.-K.Han et al., Proc. Natl. Acad. Sci. USA, 99, 14740 (2002)SKU(s): MMNP-002A, MMNP-002B
Catalog No. 50-210-9744
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.