Learn More
CPC Scientific H-Ser-Phe-Arg-Asn-Gly-Val-Gly-Ser-Gly-Val-Lys-Lys-Thr-Ser-Phe-Arg-Arg-Ala-Lys-Gln-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP012B
Encompass
This peptide is a modulator of arousal and possibly anxiety-related behaviour. It acts by activating its cognate receptor (NPSR) and inducing mobilization of intracellular Ca2+. Central administration of NPS induced wakefulness and reduced slow wave and REM sleep in rats. These peptides represent potential lead compounds for the treatment of sleep and anxiety disorders. ONE-LETTER SEQUENCE: SFRNGVGSGNKKTSFRRAKQMOLECULAR FORMULA: C95H160N34O27MOLECULAR WEIGHT:2210.53STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [412938-75-1], RESEARCH AREA: NeuropeptidesREFERENCES: Y.-L.Xu et al., Neuron, 43, 487 (2004); N.Okamura and N.K.Reinscheid, Stress, 10, 221 (2007); K.Domschke et al., Mol. Psychiatry, 16, 938 (2011)
Catalog No. 50-210-9903
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.