Learn More
CPC Scientific H-Trp-Tyr-Lys-His-Val-Ala-Ser-Pro-Arg-Tyr-His-Thr-Val-Gly-Arg-Ala-Ala-Gly-Leu-Leu-Met-Gly-Leu-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific NEUP013B
Encompass
This peptide is an endogenous ligand of the G-protein-coupled receptors GPR7 and GPR8. It dose-dependently inhibited cAMP accumulation induced by forskolin in CHO-GPR7 and CHO-GPR8 cells, with IC50 values of 0.025 and 0.178 nM, respectively. Intracerebroventricular administration of neuropeptide W-23 (human) in rats increased food intake and stimulated prolactin release.ONE-LETTER SEQUENCE: WYKHVASPRYHTVGRAAGLLMGLMOLECULAR FORMULA: C119H183N35O28S1MOLECULAR WEIGHT:2584.05STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [383415-79-0], RESEARCH AREA: NeuropeptidesREFERENCES: .Shimomura et al., J. Biol. Chem., 277, 35826 (2002); S.Brezillon et al., J. Biol. Chem., 278, 776 (2003); H.Tanaka et al., Proc. Natl. Acad. Sci. USA, 100, 6251 (2003); M.S.Mondal et al., Endocrinology, 144, 4729 (2003)
Catalog No. 50-210-9905
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.