Learn More
CPC Scientific H-Trp-Tyr-Lys-His-Val-Ala-Ser-Pro-Arg-Tyr-His-Thr-Val-Gly-Arg-Ala-Ala-Gly-Leu-Leu-Met-Gly-Leu-Arg-Arg-Ser-Pro-Tyr-Leu-Trp-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP014B
Encompass
This peptide was recently identified as the endogenous ligand for the two structurally related orphan G-protein-coupled receptors (GPCRs) GPR7 and GPR8 which are expressed in the central nervous system. NPW-30 activated and bound to both GPR7 and GPR8 at similar effective doses. Intracerebroventricular administration of NPW in rats resulted in an increased food intake and stimulation of prolactin release indicating that NPW acts as a mediator of the central control of feeding and the neuroendocrine system. ONE-LETTER SEQUENCE: WYKHVASPRYHTVGRAAGLLMGLRRSPYLWMOLECULAR FORMULA: C165H249N49O37S1MOLECULAR WEIGHT:3543.17STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [383415-80-3], RESEARCH AREA: NeuropeptidesREFERENCES: Y.Shimomura et al., J. Biol. Chem., 277, 35826 (2002); S.Brezillon et al., J. Biol. Chem., 278, 776 (2003); H.Tanaka et al., Proc. Natl. Acad. Sci. USA, 100, 6251 (2003);
Catalog No. 50-210-9907
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.