Learn More
Selleck Chemical LLC Neuropeptide Y-5mg
SDP
Supplier:  Selleck Chemical LLC P11225MG
Encompass_Preferred
Neuropeptide Y (NPY), a 36 amino acid peptide widely distributed within central nervous system neurons, is a neuroprotective agent against β-amyloid neurotoxicity which influences the synthesis and the release of nerve growth factor (NGF) by cortical neurons.
Catalog No. 50-238-9649
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.