Learn More
CPC Scientific H-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Met-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific NEUP102A
Encompass
ONE-LETTER SEQUENCE: SKPDNPGEDAPAEDMARYYSALRHYINLITRQRY-NH2MOLECULAR FORMULA: C175H269N53O54S1MOLECULAR WEIGHT:4011.45STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [150138-78-6], SYNONYMS: Neuropeptide Y (3-36) (human, rat)RESEARCH AREA: Neuropeptides
Catalog No. 50-210-9927
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.