Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Neuropeptide Y, Human and Rat, Solid, 500 ug

Catalog No. 502584653
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4653 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4653 Supplier Abcam Supplier No. AB120208500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Endogenous human and rat neuropeptide with greater than 95% purity.

  • Molecular Weight: 4,272 Da
  • Purity: >95%
  • Form: Solid
  • Composition: 100% Neuropeptide Y (human, rat)
  • CAS Number: 90880-35-6
  • Storage: Store at -20°C.
  • Shipping: Ambient - Can Ship with Ice
  • Citations: 4

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Neuropeptide Y (Human, Rat)
Substrate Properties Peptide-based Substrate
Form Solid
Content And Storage Store at -20°C.
Reactivity Human, Rat
Shipping Condition Ambient
Cross Reactivity Human, Rat
Quantity 500 μg
For Use With (Application) Sample Prep and Detection
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.