Learn More
CPC Scientific H-Tyr-Pro-Ser-Lys-Pro-Asp-Asn-Pro-Gly-Glu-Asp-Ala-Pro-Ala-Glu-Asp-Leu-Ala-Arg-Tyr-Tyr-Ser-Ala-Leu-Arg-His-Tyr-Ile-Asn-Leu-Ile-Thr-Arg-Gln-Arg-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP106B
Encompass
Brain peptide that possesses both gastrointestinal and vasoconstrictor properties.ONE-LETTER SEQUENCE: YPSKPDNPGEDAPAEDLARYYSALRHYINLITRQRY-NH2MOLECULAR FORMULA: C190H287N55O57MOLECULAR WEIGHT:4253.71STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [83589-17-7], SYNONYMS: NPY (porcine), Neuropeptide Y (porcine)RESEARCH AREA: NeuropeptidesREFERENCES: K. Tatemoto et al., PNAS, 79, 5485 (1982)
Catalog No. 50-210-9936
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.