Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neuropilin 1 Mouse anti-Human, Porcine, Clone: 2H3F6, Proteintech
SDP

Catalog No. 501736221 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6221 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6221 Supplier Proteintech Group Inc Supplier No. 600671IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Neuropilin 1 is a membrane-bound coreceptor to a tyrosine kinase receptor for both vascular endothelial growth factor and semaphorin family members. Neuropilin 1 plays versatile roles in angiogenesis, axon guidance, cell survival, migration and invasion.

Specifications

Antigen Neuropilin 1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2H3F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene NRP1
Gene Accession No. O14786
Gene Alias BDCA4, CD304, neuropilin 1, NRP, NRP1, VEGF165R
Gene Symbols NRP1
Host Species Mouse
Immunogen Neuropilin 1 Fusion Protein Ag0931
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100286859, 8829
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.