Learn More
Hamilton™ Neuros™ 1700 Series Gas Tight Syringes with Removable Needle
An industry first, the Neuros accurately dispenses 50nL to 100μL of liquid through an ultrafine needle protected by a custom-designed sleeve. The Neuros minimizes injection site damage by increasing needle rigidity and delivering a microvolume of liquid to an exact location.
Supplier:  Hamilton™ 6546021
 View more versions of this product
Catalog No. 14-785-713
Only null left
Add to Cart
Description
Description
  • Needle rigidity improves insertion path accuracy
  • Minimal tissue damage reduces injection variability
  • Reduced sample loss saves money and materials
  • Fine gauge needle creates smaller injection sites
  • Compatibility with most infusion pumps and stereotaxic holders means an easy integration into existing processes
Specifications
Specifications
Borosilicate Glass, PTFE (Plunger)
0.108 mm
7.75 mm
Controlled animal injections
Gastight
239°F
50°F
4
33 G
0.145 mm
0.210 mm
0 to 20 mm
1000 psig (68.9 bar)
115°C
1710 RN
100 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.