Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech Neuroserpin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. p-7245642
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P271BT25 25 μg
500P271BT50 50 μg
50-271-8548 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P271BT25 Supplier PeproTech Supplier No. 500P271BT25UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MTGATFPEEA IADLSVNMYN RLRATGEDEN ILFSPLSIAL AMGMMELGAQ GSTQKEIRHS MGYDSLKNGE EFSFLKEFSN MVTAKESQYV MKIANSLFVQ NGFHVNEEFL QMMKKYFNAA VNHVDFSQNV AVANYINKWV ENNTNNLVKD LVSPRDFDAA TYLALINAVY FKGNWKSQFR PENTRTFSFT KDDESEVQIP MMYQQGEFYY GEFSDGSNEA GGIYQVLEIP YEGDEISMML VLSRQEVPLA TLEPLVKAQL VEEWANSVKK QKVEVYLPRF TVEQEIDLKD VLKALGITEI FIKDANLTGL SDNKEIFLSK AIHKSFLEVN EEGSEAAAVS GMIAISRMAV LYPQVIVDHP FFFLIRNRRT GTILFMGRVM HPETMNTSGH DFEEL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Neuroserpin. Anti-Human Neuroserpin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Neuroserpin by sandwich ELISA (using 100 mL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti- Human Neuroserpin (500-P271) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Neuroserpin. Western Blot: To detect Human Neuroserpin by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 mg/mL. When used in conjunction with compatible development reagents the detection limit for Recombinant Human Neuroserpin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P271BT-1MG will be provided as 2 x 500 μg

This gene encodes a member of the serpin superfamily of serine proteinase inhibitors. The protein is primarily secreted by axons in the brain, and preferentially reacts with and inhibits tissue-type plasminogen activator. It is thought to play a role in the regulation of axonal growth and the development of synaptic plasticity. Mutations in this gene result in familial encephalopathy with neuroserpin inclusion bodies (FENIB), which is a dominantly inherited form of familial encephalopathy and epilepsy characterized by the accumulation of mutant neuroserpin polymers. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neuroserpin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene Serpini1
Gene Accession No. Q99574
Gene Alias AI837402; DKFZp781N13156; neuroserpin; Ns; Peptidase inhibitor 12; PI12; PI-12; protease inhibitor 17; raPIT5a; serine (or cysteine) peptidase inhibitor, clade I, member 1; serine (or cysteine) proteinase inhibitor clade member 1; serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1; serine protease inhibitor 17; serpin family I member 1; serpin I1; serpin peptidase inhibitor clade I member 1; serpin peptidase inhibitor, clade I (neuroserpin), member 1; serpin peptidase inhibitor, clade I, member 1; SERPINI1; Spi17
Gene Symbols Serpini1
Host Species Rabbit
Immunogen E.coli-derived, 44.8 kDa Recombinant Human Neuroserpin.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5274
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.