Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NeuroTACS In Situ Apoptosis Detection Kit
GSA_VASDP

Catalog No. 482330K
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
482330K 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 482330K Supplier R&D Systems™ Supplier No. 482330K

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Proteinase K, 30uL; NeuroPore™, 5mL; TdT Enzyme, 30uL; 50X Mn+2, 30uL; Blue counterstain, 50mL; TACS-Nuclease and buffer; dNTP mix and buffers for TdT, DAB solution and enhancer

Designed for cells or tissue sections from the nervous system, and is compatible with immunocytochemistry to identify cell types and antigens by using cell specific antibodies

Primary neuronal cell cultures are often maintained in mixed culture, therefore, the ability to identify specific cells using criterion other than morphology is important. Similarly, it can be difficult to identify cell types within central nervous system

Complete kits contain Streptavidin-HRP, DAB, Blue Counterstain, and reagents necessary for generating positive controls
Study fixed cells, embedded tissues and frozen tissues using light microscopy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 30 Tests
Content And Storage The product requires storage at -20 to -70° C, 2 - 8° C, and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.