Learn More
CPC Scientific Glp-Leu-Tyr-Glu-Asn-Lys-Pro-Arg-Arg-Pro-Tyr-Ile-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP180A
Encompass
Peptide isolated primarily from the nervous system of the hypothalamus, median eminence, substantial nigra and brain stem that has many neurotransmitter-like characteristics. It can lower blood pressure, increase vascular permeability and increase plasma levels of glucagon, glucose, growth hormone, and porlactin in rats. ONE-LETTER SEQUENCE: Glp-LYENKPRRPYILMOLECULAR FORMULA: C78H121N21O20MOLECULAR WEIGHT:1672.95STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [55508-42-4], RESEARCH AREA: NeuropeptidesREFERENCES: R.E. Carraway et al., J. Biol. Chem., 248, 6854 (1973); J.E. Rivier et al., J. Med. Chem., 20, 1409 (1977)
Catalog No. 50-210-9958
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.