Learn More
R&D Systems™ NeuroXVivo Rat Motor Neuron Culture Kit
Description
Key Benefits:
- Features an optimized serum-free media for rat motor neuron cultures
- Utilizes R&D Systems™ Recombinant BDNF, CNTF, GDNF, and Laminin α4
- Detailed protocol is included
- Ideal for both non-experienced and experienced neural cell culture researchers
Kit Contents:
This kit contains the following reagents to maintain mouse pluripotent stem cells in culture:
- Recombinant BDNF (1000X)
- Recombinant CNTF (1000X)
- Recombinant GDNF (1000X)
- Recombinant Laminin α4 (1000X)
- Motor Neuron Culture Media Supplement (10X)
- Reconstitution Buffer 1
Limitations:
- FOR LABORATORY RESEARCH USE ONLY. NOT FOR USE IN DIAGNOSTIC PROCEDURES.
- The safety and efficacy of this product in diagnostic or other clinical uses has not been established.
- This reagent should not be used beyond the expiration date indicated on the label.
- Results may vary due to variations among cells derived from different donors.
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.