Learn More
StatLab™ NeutraGreen™ Liquid Neutralizer



Description
- Comes in liquid form with no powders to mix or dissolve
- Waste reaction time of 2 to 3 hr enables same day disposal
- Third-party environmental data confirms NeutraGreen™ will neutralize waste formalin to a material that is non-toxic and non-hazardous
- Test with NeutraGreen™ Test Strips to ensure waste has been properly neutralized for drain disposal
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity (English) | 2.5 gal. |
| Capacity (Metric) | 9.46 L |
| Material | Pre-treated Formalin Neutralizer |
| Type | Formalin Neutralizer |
| Format | Cube |
| For Use With (Application) | Neutralizing Waste Formalin |
| Packaging Quantity | 2.5 gal. |
| Weight (English) | 26 lb. |
| Weight (Metric) | 11.79 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.