Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Neutral Litmus Test Paper
SCI_ED

Catalog No. S80849 Shop All Eisco Products
Change view
Click to view available options
pH Range:
0 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
S80849 0 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80849 Supplier Eisco™ Supplier No. NEULITPK12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Qualitatively indicate if a solution is neutral, acidic, or basic without providing a specific pH value for the solution

  • Tests the neutrality, acidity or alkalinity of solution
  • Does not include a color chart
  • Will not provide a specific pH value of the solution
  • Test strip turns red in the presence of an acidic solution (pH < 7)
  • Test strip turns blue in the presence of an alkaline solution (pH ˃ 7)
  • 100 Strips/Vial, 12 Vials/Pack

Specifications

Color Indicator Red (Acid), Blue (Base)
pH Range 0 to 14
For Use With (Application) Detection of a Neutral, Acidic, or Basic Solution
Type Litmus Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.