Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neutral Red, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18611929 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
10 g
25 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-611-929 1 kg
18-611-930 10 g
18-611-931 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-611-929 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. N34451KGBL
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Neutral Red
CAS 553-24-2
CAS Max % 1
Grade ACS
Packaging Amber Glass Bottle
Color Dark Green
Quantity 1 kg

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.