Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scigen Neutralex™ Neutra-Wipes™ – Refill Rolls

Catalog No. 23730575 Shop All Scigen Inc Products
Change view
Click to view available options
Packaging Quantity:
1 Roll, 60 Wipes
4 rolls, 240 Wipes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
23-730-575 4 rolls, 240 Wipes
23-730-576 1 Roll, 60 Wipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-730-575 Supplier Scigen Supplier No. 4279
Add to Cart
Add to Cart

Includes

4 rolls only (no dispenser).

Neutra-Wipes™ rapidly neutralize small formalin spills and absorb 30 to 50 mL of liquid. Neutralex™ products are certified by the California Environmental Protection Agency. The seal indicated that the formalin treatment process results in non-hazardous drain safe waste.

  • Absorbs and neutralizes noxious vapors from small formalin and aldehyde spills
  • 4 rolls of 60 wipes (12 x 12.5 in.) per roll
  • Manufactured using ISO 13485 quality system
  • Ideal as liners on storage carts, shelves, and in rooms where formalin and aldehyde are kept or transported—neutralizing any spillage and helping to eliminate noxious vapors
  • Designed specifically for use with other Neutralex products for maximum effectiveness in treating formalin and aldehyde waste
  • After wiping up a formalin or aldehyde spill, simply dispose of the wipe in an appropriate disposal container – the liquid has been properly neutralized

Note: For convenient pop-up dispenser, see Neutra-Wipes (4278) which includes dispenser and 1 roll of wipes.

Specifications

Color White
Description Instantaneously neutralize formalin, while absorbing harmful liquids
Capacity (English) 1 to 1.69 oz.
Capacity (Metric) 30 to 50 mL
Material Heavy-duty Cloth
Type Absorbent Wipes
For Use With (Application) Small Formalin Spills
Includes 4 rolls (no dispenser).
Packaging Quantity 4 rolls, 240 Wipes
Dimensions (L x W) 12 x 12.5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.