Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab NeutraSafe Aldehyde Test Kit
SDP

Catalog No. 22046643
Change view
Click to view available options
Packaging:
25 Tests/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
22-046-643 25 Tests/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-046-643 Supplier StatLab Supplier No. FTK/25
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Test kit contains collection vessel, reagent powder packet, and color indicator solution.

  • Kit contains collection vessel, reagent powder packet, and color indicator solution
  • 25 tests per kit
  • For use with NeutraSafe powdered aldehyde

Specifications

Sample Type Swab
Type Aldehyde Test Kit
Packaging 25 Tests/Kit
Quantity 25/Pk.
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.