Learn More
Getinge USA Neutral Instrument Detergent NeutraWash Liquid Concentrate 4x1 gal. Jug Mild Scent
Supplier:  Getinge USA 61301605274
Encompass
Special scale and mineral dissolving agents actually brighten metal surfaces and extend the life of instruments and equipment. Incorporated rinse aid eliminates spotting and filming.
Catalog No. NC1840913
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.