Learn More
Labconco Neutrodine Unisorb EXT
SDP
Catalog No. 13000016 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector Echo Filtered Hood
Catalog No. For Use With (Equipment)
13-000-016 Protector Echo Filtered Hood
1 options
Catalog No. 13-000-016 Supplier Labconco Supplier No. 9577460
Add to Cart
Add to Cart
Unisorb Molecular Filter unit for multi-purpose use of acids/bases/solvents
  • Neutrodine Unisorb molecular carbon filter unit for multi-purpose use of acids/bases/solvents, separated or combined
  • For use with Protector Echo and Airo Filtered Fume Hoods manufactured after February 2025
  • Neutrodine Unisorb Molecular Filter (external connector)
Specifications
For Use With (Equipment) Protector Echo Filtered Hood
Product Type Carbon Filter