Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Neutrophil Elastase/ELA2 Recombinant Protein Antigen
SDP

Catalog No. RD10183776 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10183776 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10183776 Supplier R&D Systems™ Supplier No. NBP325010PEP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Recombinant Protein

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 1991
Quantity 100 μL
Storage Requirements Store at -20C. Avoid freeze-thaw cycles.
Gene Alias Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Gene Symbol ELANE
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP3-25010It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml.For current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com.
Research Category Cancer, Neuroscience
Product Line Novus
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Neutrophil Elastase/ELA2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.