Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neutrophil elastase Monoclonal Antibody (ELA10-103.2), Invitrogen™

Catalog No. p-7245250
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546148 100 μg
01-671-562 500 μg
PIMA546009 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546148 Supplier Invitrogen™ Supplier No. MA546148
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10609 detects Neutrophil elastase from human samples. MA1-10609 has been successfully used in ELISA applications. The MA1-10609 immunogen is purified human Neutrophil Elastase.

The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neutrophil elastase
Applications ELISA
Classification Monoclonal
Clone ELA10-103.2
Concentration 1.28 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ELANE
Gene Accession No. P08246
Gene Alias bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease
Gene Symbols ELANE
Host Species Mouse
Immunogen Purified human Neutrophil Elastase.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1991
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.