Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Neutrophil elastase Monoclonal Antibody (ELA11-207.3), Invitrogen™

Mouse Monoclonal Antibody
$450.00 - $1108.00
Specifications
| Antigen | Neutrophil elastase |
|---|---|
| Clone | ELA11-207.3 |
| Concentration | 0.91 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA546144
|
Invitrogen™
MA546144 |
100 μg |
Each for $450.00
|
|
|||||
01-671-560
![]() |
Invitrogen™
MA110607 |
500 μg |
Each for $600.50
|
|
|||||
|
PIMA546005
|
Invitrogen™
MA546005 |
1 mg |
Each for $1,108.00
|
|
|||||
Description
MA1-10607 detects Neutrophil elastase from human samples. MA1-10607 has been successfully used in ELISA applications. The MA1-10607 immunogen is purified human Neutrophil Elastase.
The neutrophil form of elastase is 218 amino acids long, with two asparagine-linked carbohydrate chains (see glycosylation). It is present in azurophil granules in the neutrophil cytoplasm. There appear to be two forms of neutrophil elastase, termed IIa and IIb.Specifications
| Neutrophil elastase | |
| 0.91 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| bone marrow serine protease; ELA2; ELA2A; ELANE; Elastase; elastase 2, neutrophil; elastase, neutrophil expressed; Elastase2; elastase-2; F430011M15Rik; GE; granulocyte-derived elastase; HLE; HNE; Human leukocyte elastase; Leukocyte elastase; medullasin; NE; neutrophil elastase; PE-1; PMN elastase; PMN-E; polymorphonuclear elastase; SCN1; serine protease | |
| ELANE | |
| IgM | |
| Protein A | |
| Antibody |
| ELA11-207.3 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P08246 | |
| 1991 | |
| Purified human Neutrophil Elastase. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| ELANE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title