Learn More
AdipoGen anti-Neutrophils (mouse), mAb (blocking) (Nimp-R14) (100 µg)
SDP
Supplier:  AdipoGen AG20B0043C100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse neutrophils (surface protein mouse Ly-6G) and may recognize Ly-6Chigh cells (inflammatory monocytes). Isotype: Rat IgG2b. Clone: Nimp-R14. Applications: FACS, FUNC (Blocking), ICC, IHC. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Neutrophils are key components of the inflammatory response and as such contribute to the killing of microorganisms. In addition, recent evidence suggests their involvement in the development of the immune response. The Nimp-R14 mAb displays features of an optimal reagent allowing the selective depletion of mouse neutrophils in vivo.
Catalog No. 50-205-2951
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.