Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ New Brunswick Innova™ 42/42 R - Stackable Incubator Shaker PROMO

Catalog No. 05400162PM Shop All Eppendorf North America Products
Change view
Click to view available options
Orbit:
19 mm
25 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Orbit
05-400-162PM 25 mm
05-400-157PM 19 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-400-162PM Supplier Eppendorf™ Supplier No. 2231001124

missing translation for 'validMainframeMsg'

Versatile and reliable. Stackable, can be placed on or under the bench, and come with a static shelf to be used to incubate agar plates while shaking cultures in flasks (Optional: static culture drawer, gassing manifold, remote alarm, and humidity monitor and drip pan.

  • High capacity in a small footprint: for example, 4 x 5 L or 2 x 6 L flasks on universal platform; device footprint 4731 cm2 /733 in.2 (63.5 cm/25 in. width, 74.5 cm/29.3 in. depth)
  • Ergonomical and safe interface handling: Top mounted controls and display make bending down unnecessary during handling, avoids accidental parameter changes, and can be reached easily even in a double stack
  • Easy to clean: Spill diverter and drainable built-in spill pan with rounded corners protect the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation); airflow UV-germicidal light optional
  • Lab position flexibility: Lab position on or under bench, 2x stackable
  • Parallel static cultivation: One height adjustable static shelf included, optional culture drawer and additional shelves available
  • Sample evaporation protection: Bottom spill pan usable as water reservoir; optional culture drawer, for Agar plates, for example
  • Visual sample control: Large viewing window and internal light provide easy viewing of samples without the need to open the door
  • Reliable 24/7 with high loads: Proven and tested Innova triple eccentric counterbalanced drive (three weight-supporting shafts), max. load 15 kg
  • Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
  • Options and Accessories: Orbit 19 or 25 mm, photosynthetic light bank, UV germicidal light, culture drawer for evaporation protection of Agar plates, additional static shelves (one included), humidity monitoring, remote monitoring with VisioNize™ Lab Suite and more
  • Speed: 25 to 400 rpm, 25 to 300 rpm when using flasks >500 mL in stacked devices
  • Certifications/Compliance: 2014/35/EU: EN 61010-1, EN 61010-2-010, EN 61010-2-051, 2014/30/EU: EN 61326-1, EN 55011 (Class A), 2011/65/EU: EN IEC 63000, IEC 61010-1, IEC 61010-2-010, IEC 61010-2-051, UL 61010-1, UL 61010-2-010, UL 61010-2-051, CAN/CSA C22.2 No. 61010-1-12, CSA C22.2 No. 61010-2-010, CSA C22.2 No. 61010-2-051, IEC 61326-1, CISPR 11, FCC 47 CFR Part 15 (Class A)
TRUSTED_SUSTAINABILITY

Specifications

Orbit 25 mm
Load Bearing Capacity (Metric) 9 to 15.8 kg
Speed Range 25 to 400 rpm
Electrical Requirements 120 V 50/60 Hz
Voltage 120 V
Temperature Range (Metric) 20°C Below Ambient to 80°C
Amperage 1500 VA (12.5 A)
Holds 6 L Flasks
Product Type Incubator Shaker
Load Bearing Capacity (English) 20 to 35 lb.
Temperature Range (English) 68°F below ambient to 176°F
Temperature Uniformity ±0.25°C at 37°C
Frequency 60 Hz
Material Aluminum Platform
Depth (English) 29.3 in.
Depth (English) Interior 20.3 in.
Depth (Metric) 74.5 cm
Depth (Metric) Interior 51.5 cm
Dimensions (D x W x H) Interior 51.5 x 51.7 x 47.7 cm (20.3 x 20.4 x 18.8 in.)
Dimensions (LxW) Platform 46 x 46 cm (18 x 18 in.)
Dimensions (L x W x H) 63.5 x 74.5 x 81.8 cm / 25 x 29.3 x 32.8 in.
Height (English) 32.2 in.
Height (English) Interior 18.8 in.
Height (Metric) 81.8 cm
Height (Metric) Interior 47.7 cm
Width (English) 25 in.
Width (English) Interior 20.4 in.
Width (Metric) 63.5 cm
Width (Metric) Interior 51.7 cm
Weight (English) 266 lb.
Weight (Metric) 121 kg
Accuracy ±1 rpm
Model Innova 42R
Display LED Digital Electric Display
For Use With (Application) Plasmid DNA Production and Protein Expression, Biofuel Research, Grow Bacteria, Yeast, Fungus, Photosynthetic Organisms
Humidity Range 20 to 80%, Non-condensing
Includes Accessories
Interface USB, Ethernet
Platform Style Universal/Dedicated
Style Universal/Dedicated
Timer Range 0.01 sec. to 95 hr. 59 min., Continuous
Warranty and Service Offering 3 Year
Refrigerated Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.