Learn More
Eppendorf™ Scientific Innova™ Shakers::Accessories for Innova™ Platform Shakers::Capacity Upgrade Kits
Catalog No. 14278136 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Innova 2100
Catalog No. For Use With (Equipment)
14-278-136 Innova 2100
1 options
This item is not returnable. View return policy
Catalog No. 14-278-136 Supplier Eppendorf™ Supplier No. M11949926
This item is not returnable. View return policy
Includes everything needed for conversion of Model 2100 to Model 2150
Convert Model 2000 for 12L × 16 in.W (30.5 × 41cm) platform or Model 2100 for 18L × 24 in.W (46 × 61cm) platform. Include everything needed for conversion; without platform. Must be installed by a qualified service person; contact your Fisher Scientific™ Customer Service Representative for assistance.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Innova 2100