Learn More
Eppendorf™ Scientific shaker Microplate Platform
Catalog No. 1427832 Shop All Eppendorf North America Products
Change view
Click to view available options
No. of Positions:
5
Catalog No. No. of Positions
14-278-32 5
1 options
Catalog No. 14-278-32 Supplier Eppendorf™ Supplier No. TTR221
Add to Cart
Add to Cart
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Mount on universal platforms
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microplate Rack
For Use With (Equipment) Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series
No. of Positions 5