Learn More
Eppendorf™ Universal Platform for Model C76 Water Bath Shaker
Catalog No. 14278031 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Model C76 water bath shaker
Catalog No. For Use With (Equipment)
14-278-031 Model C76 water bath shaker
1 options
Catalog No. 14-278-031 Supplier Eppendorf™ Supplier No. M12319930
Add to Cart
Add to Cart
Both universal platforms and dedicated platforms are available in a wide variety of material and size perfect for virtually every requirement. Universal platforms have multiple holes allowing you to use virtually any combination of clamps and other accessories. Dedicate platforms are used when just one flask size will be used.
  • Holds different combinations of flask clamps
  • Maximum number of flasks: 82 (10mL), 16 (25mL), 14 (50 or 125mL), 8 (250mL), 6 (500mL), 4 (1L), or 2 (2L)
  • Measures 16.5W × 12 in.L (42 × 30cm)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Model C76 water bath shaker
Type Universal Platform
Dimensions (L x W) 30.5 x 41.9 cm