Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig High-Visibility Yellow Mats and Rolls
SDP

Catalog No. 189995700 Shop All New Pig Corporation Products
Change view
Click to view available options
Capacity (English):
28.16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
18-999-5700 28.16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-5700 Supplier New Pig Supplier No. MAT605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hard-to-miss hi-viz yellow mat that draws attention to spill cleanups.

  • Heavyweight mat pads are ideal for soaking up big spills and heavy leaks around machines and equipment; dispenser box fits neatly on shelves or carts
  • Bright yellow color stands out so it's easy for workers to see hazards, repairs or cleanups in progress, even in poorly lit areas.
  • PIG Mat lasts 2 times longer than competitive mats and won't rip, tear or fray even when saturated; liquids absorb quickly for faster, easier cleanups and safer workplaces
  • Absorbs most common industrial liquids - oils, coolants, solvents, water and more

Specifications

Color Yellow
Capacity (English) 28.16 oz.
Capacity (Metric) 0.83 L
Material Polypropylene
Type High Visibility Absorbent Mat Pad
For Use With (Application) Absorbs oils, coolants, solvents and water
Dimensions (L x W) 20 x 15 in. (50.8 x 38 cm)
Length (English) 20 in.
Length (Metric) 50.8 cm
Width (English) 15 in.
Width (Metric) 38 cm
Weight (English) 18 lb.
Weight (Metric) 8.16 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.