Description
*These services can only be ordered in combination with a NEW Eppendorf Pipette. Ensure that an Eppendorf pipette/dispenser with a matching number of channels is also added to the order.
- The pipette will be calibrated in the local Eppendorf calibration facility before delivery.
ISO 17025 Calibration:
- Calibration test of 3 volumes, 10 measurements per volume for a single channel pipette/dispenser.
- Calibration process and documentation are ISO 17025 compliant.
- Issuing of calibration certificate with expanded measurement uncertainties and calibration label.
ISO 8655 Test:
- Routine test for 3 volumes, 10 measurements per volume.
- Issuing of test report without expanded measurement uncertainties and test documentation label.
3x4 Measurements Test:
- Routine test for 3 volumes, 4 measurements per volume.
- Issuing of test report without expanded measurement uncertainties and test documentation label.
Specifications
Specifications
| For Use With (Equipment) | Eppendorf 8-channel Pipettes |
| Description | Includes ISO 17025 compliant calibration certificate with expanded measurement uncertainties and calibration label |
| Calibration Points | 3 x 10 Measurements/Volume |
| Duration | 12 Months |
| Calibration Type | ISO 17025 Compliant Calibration and documentation |
| Product Type | Service Plan |
| Service Offering | Calibration Test |