Description
The New Standard™ includes a warmer ready base, 100 micron 3-axes, left-hand manipulator arm, separate rat and mouse adaptors, dual-sided (18° and 45°) rat ear bars, triple point mouse ear bars, and corner clamp probe holder. Available as dual, ultra-precise, digital or motorized
Purchase Stoelting’s Rodent Warmer Control Box X1 (item 53800), or Rodent Warmer Control Box X2 (item 53850) to use the warming function. Like the classic Lab Standard™, the new Standard Stereotaxic Instrument has larger laser engraved vernier scales (accurate to 100 um), 80 mm of ventral-dorsal, medial-lateral and anterior-posterior travel, absolute lock at 90° ventral-dorsal and brass-bushings for electrical grounding. It is available as a digital or motorized model.
The New Standard™ design is what sets it apart:
- Revolutionary vertically-adjusting ear and nose bar posts
- Revolutionary horizontally-adjusting ear and nose bar posts
- Designed for both Rats and Mice all on the same base
- No U-frame, maximizes operating space
- Traditional and non-traumatic Rat ear bars
- Includes the innovative Mouse ear bars
- Includes Mouse and Rat Nose adaptors
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Animal | Mouse, Rat |
| Model | Ultra-Precise |