Learn More
Stoelting™ New Standard™ Stereotaxic Instruments for Rats and Mice
SDP
Catalog No. 10000390 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Base only
Digital
Digital, Ultra-Precise
Dual
Dual, Digital
Motorized
Standard Base
Ultra-Precise
Ultra-Precise, Dual
Ultra-Precise, Dual, Digital
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-390 Digital, Ultra-Precise
10-000-387 Digital
10-000-131 Dual
10-000-391 Dual, Digital
10-000-393 Ultra-Precise, Dual, Digital
10-000-388 Motorized
10-000-129 Standard Base
10-000-389 Ultra-Precise
10-000-392 Ultra-Precise, Dual
10-000-130 Base only
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 10-000-390 Supplier Stoelting™ Supplier No. 51500UD
Add to Cart
Add to Cart
Request a Quote
The innovative design of the New Standard™ Stereotaxic Instrument provides users with the ability to perform surgeries on both Mice and Rats on the same base.
The New Standard™ includes a warmer ready base, 100 micron 3-axes, left-hand manipulator arm, separate rat and mouse adaptors, dual-sided (18° and 45°) rat ear bars, triple point mouse ear bars, and corner clamp probe holder. Available as dual, ultra-precise, digital or motorized
Purchase Stoelting’s Rodent Warmer Control Box X1 (item 53800), or Rodent Warmer Control Box X2 (item 53850) to use the warming function. Like the classic Lab Standard™, the new Standard Stereotaxic Instrument has larger laser engraved vernier scales (accurate to 100 um), 80 mm of ventral-dorsal, medial-lateral and anterior-posterior travel, absolute lock at 90° ventral-dorsal and brass-bushings for electrical grounding. It is available as a digital or motorized model.
The New Standard™ design is what sets it apart:
  • Revolutionary vertically-adjusting ear and nose bar posts
  • Revolutionary horizontally-adjusting ear and nose bar posts
  • Designed for both Rats and Mice all on the same base
  • No U-frame, maximizes operating space
  • Traditional and non-traumatic Rat ear bars
  • Includes the innovative Mouse ear bars
  • Includes Mouse and Rat Nose adaptors
Specifications
For Use With (Application) Neuroscience
Animal Mouse, Rat
Model Digital, Ultra-Precise