Learn More
Specifications
Specifications
| Color | Black |
| Quantity | 1 Ea. |
| Includes | Lock, clear panels |
| For Use With (Application) | For security conscious facilities such as airports, train stations, sports facilities, government and office complexes and convention centres |
| Material | Polycarbonate |
| Product Type | Security Container |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.