Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubbermaid™ Steel Sponge Mop with Cellulose Head

Catalog No. 19065270 Shop All Newell Rubbermaid Inc Products
Change view
Click to view available options
Color:
Bronze
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-065-270 Bronze
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-065-270 Supplier Rubbermaid™ Supplier No. FG643500BRNZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Sponge Mop
Color Bronze
Length (English) 12 in.
Includes Steel Roller Sponge Mop with Cellulose Head
For Use With (Application) For Easy Cleaning
Material Steel, Cellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.