Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubbermaid™ Super Stitch™ Cotton Looped End Wet Mops

Catalog No. 19046803 Shop All Newell Rubbermaid Inc Products
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-803 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-046-803 Supplier Rubbermaid™ Supplier No. FGD15306WH00
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Wet Mop
Color White
Length (English) 35 to 41 in.
Width (English) 5.5 to 6.5 in.
Size Large
Includes Mop
For Use With (Application) For General Purpose
Material Cotton
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.