Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rubbermaid™ Universal Headband Cotton Mops

Catalog No. 19120408 Shop All Newell Rubbermaid Inc Products
Change view
Click to view available options
Capacity:
24 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
19-120-408 24 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-120-408 Supplier Rubbermaid™ Supplier No. FGE13800WH00
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Product Type Headband Mop
Color White
Capacity 24 oz.
Quantity 12/Cs.
For Use With (Application) For General Cleaning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.