Learn More
Description
Complete coverage of 15 concepts: Whole Numbers, Patterns and Algebra, Mass, Addition and Subtraction, Data, Time, Multiplication and Division, Length, Three-Dimensional Space, Chance, Volume and Capacity, Position
- Developed by an experienced educator and classroom-tested for more than a decade
- Wealth of challenging, mathematical exercises focused on leveled, standards-based topics
- Fun-filled means of motivating students to think more deeply, investigate, explain, and understand problem-solving strategies
- Concepts divided into two sections: one per semester
- Each section is two to three pages in length and followed by an assessment for immediate and continuous feedback
Specifications
Specifications
| Grade Level | Elementary |
| Grade | 1 |
| Format | E-books/Workbooks |
| Product Type | I Love Hands-on Math Reproducible Workbook |
| For Use With (Application) | Fun filled means of motivating students to think more deeply, investigate, explain and understand problem-solving strategies |
| Subject | Mathematics |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
