Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Physical Science Curriculum Mastery™ Posters
SCI_ED

Catalog No. s04383 Shop All NewPath Learning Products
Change view
Click to view available options
Product Type:
Electricity Poster
Forces and Motion Poster
Light Poster
Sound Poster
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S04383 Forces and Motion Poster
S04384 Electricity Poster
S04387 Light Poster
S04386 Sound Poster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S04383 Supplier NewPath Learning Supplier No. 346801
Add to Cart
Add to Cart

Graphic overview of key physical science topics.

  • Provide a comprehensive look at key, standards-based topics
  • Laminated
  • Grades 6 to 10

Forces & Motion Poster:
Provides a detailed overview of force, gravity, motion and momentum as well as Newton's Laws

Electricity Poster:
A graphic look at electric charge, current, circuits and power! Also provides an overview of voltage, resistance and Ohm's Law

Magnetism Poster:
What is a magnet? How do magnetic fields work? How can magnetism generate electricity?

Sound Poster:
Take a comprehensive look at sound waves as well as their characteristics, properties and uses

Light Poster:
Features a detailed overview of light waves; reflection and refraction; and mirrors, lenses and their applications

Order Info

Size: 23 x 35 in. (58.4 x 89cm).

Specifications

Product Type Forces and Motion Poster
For Use With (Application) Provides a detailed overview of forces, gravity, motion and momentum as well as Newton's law
Certifications/Compliance Academic Standard for California, Florida, National, New York, Texas
Grade Level Intermediate - Middle School, High School
Grade 6 to 12
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.