Learn More
United Scientific Supplies Newton's Ring Apparatus
SDPSCI_ED
Catalog No. S27835 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Application):
Measuring Newton's Rings
Catalog No. For Use With (Application)
S27835 Measuring Newton's Rings
1 options
Catalog No. S27835 Supplier United Scientific Supplies Supplier No. NEWRIN2
Add to Cart
Add to Cart
Consists of a plano-convex lens atop a flat glass plate, all secured by a plastic frame
  • Thumbscrews allow for adjustments to pressure between lens and plate
Specifications
Product Type Newton's Rings
For Use With (Application) Measuring Newton's Rings