Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nexilin Polyclonal Antibody

Catalog No. PIPA5121743
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121743 20 μL
PIPA5119307 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121743 Supplier Invitrogen™ Supplier No. PA5121743
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Nexilin Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Involved in regulating cell migration through association with the actin cytoskeleton. Has an essential role in the maintenance of Z line and sarcomere integrity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nexilin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene NEXN
Gene Accession No. Q7TPW1, Q9Z2J4
Gene Alias 1110046H09Rik; AA553326; CMH20; F-actin binding protein; F-actin-binding protein; Nelin; Nexilin; nexilin (F actin binding protein); nexilin alpha; nexilin beta; nexilin F-actin binding protein; nexilin gamma; NEXN; nexn {ECO:0000312; nF actin binding protein; RGD:708354}
Gene Symbols NEXN
Host Species Rabbit
Immunogen Recombinant protein Nexilin. The antigen corresponds to amino acid range 214-455 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246172, 68810
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.