Learn More
TARGETMOL CHEMICALS INC NEXOPAMIL 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T28164
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Nexopamil (LU-49938) is a calcium channel antagonist and a 5-hydroxytryptamine 2A receptor antagonist. Nexopamil inhibits 5-HT-induced contraction and proliferation of mesenchymal cells. purity: 99%
Catalog No. 50-258-0541
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.