Learn More
Nextteq™ Gastec™ GV100S Air Sampling Kit
SDP
Catalog No. 19026372 Shop All Nextteq Products
Change view
Click to view available options
Product Type:
Air Sampling Pump
Catalog No. Product Type
19-026-372 Air Sampling Pump
1 options
Catalog No. 19-026-372 Supplier Nextteq™ Supplier No. GV100STR
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Accurately, efficiently measures more than 500 gases and vapors over a wide range of environmental conditions; Produces clear, easy-to-read results
This precision piston pump uses glass detector tubes to accurately and quickly measure over 500 gases and vapors. Each tube contains a reagent that is specifically sensitive to a particular gas or vapor. Break off ends of tube, insert in handheld pump, and pull pump handle out. A precisely measured volume of ambient air is drawn into the tube, reacts with the reagent, and produces a stain that can be read against the calibrations on the tube, like a thermometer. The longer the stain, the higher the concentration.
The springless pump operates on vacuum principle for long-lasting accuracy. Fail-safe handle locks precisely at 50 or 100cc for accurate sample size. A flow finish indicator, conspicuously located in the pump handle, indicates when the tube has completed its reading. High-impact-resistant exterior with synthetic rubber sheath, metal alloy internal cylinder. Features built-in tube tip breaker, carrying case, spare parts, and instructions.
Warranty and Services
Lifetime warranty.
Specifications
Includes Built-in Tube Tip Breaker, Carry Case, Instructions
For Use With (Application) Efficiently measures more than 500 gases and vapors over a wide range of environmental conditions
Product Type Air Sampling Pump