Learn More
Labconco Nexus Remote Communication Board
GSA_VASDP
Catalog No. 17200209 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Enclosures
Catalog No. For Use With (Equipment)
17-200-209 Enclosures
1 options
Catalog No. 17-200-209 Supplier Labconco Supplier No. 5321400
Add to Cart
Add to Cart
Nexus Remote Communication Board kit includes contacts required for remote system management.
  • Kit includes three dry contact relays
  • USB data output
  • RS-232 RJ45 data output
  • Ethernet pass-through for easy connection
  • Mounts on the back of the Nexus Horizontal Clean Bench
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) Enclosures
Product Type Remote Communication Board