Learn More
Medchemexpress LLC HY-130246 5mg Medchemexpress, NF-56-EJ40 CAS:2380230-73-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1302465MG
Encompass_Preferred
Medchemexpress, HY-130246 5mg NF-56-EJ40 CAS:2380230-73-7 NF-56-EJ40 is a potent, high-affinity, and highly selective human SUCNR1 (GPR91) antagonist with an IC50 of 25 nM and a Ki of 33 nM, and shows almost no activity towards rat SUCNR1. NF-56-EJ40 has high affinity for humanized rat SUCNR1 with a Ki value of 17.4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9824
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.